accessibility menu, dialog, popup

UserName

H3COOS

CAS RN 7605-25-6

IUPAC Name: ethyl 2-(phenylsulfanyl)acetate
Synonyms: ethyl 2-(phenylsulfanyl)acetate
Molecular Weight (g/mol): 196.26
Molecular Formula: C10H12O2S
InChi Key: SEDRTXNDGKRHBL-UHFFFAOYSA-N
SMILES: CCOC(=O)CSC1=CC=CC=C1
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)