missing translation for 'onlineSavingsMsg'
Learn More

ethyle2-(phenylthio)acetate, 97%, Thermo Scientific™

Product Code. 10794003
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10794003 10 g 10g
10272692 1 g 1g
10496852 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10794003 Supplier Thermo Scientific Maybridge Supplier No. KM08693EA
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 7605-25-6
Molecular Formula C10H12O2S
Molecular Weight (g/mol) 196.264
MDL Number MFCD00145107
InChI Key SEDRTXNDGKRHBL-UHFFFAOYSA-N
Synonym ethyl phenylthio acetate, ethyl 2-phenylthio acetate, ethyl 2-phenylsulfanyl acetate, acetic acid, phenylthio-, ethyl ester, ethyl 2-phenylthioacetate, phenylthio acetic acid ethyl ester, phenylthio acetic acid, ethyl ester, ethyl phenylsulfanyl acetate, pubchem10853
PubChem CID 97312
IUPAC Name ethyl 2-phenylsulfanylacetate
SMILES CCOC(=O)CSC1=CC=CC=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Linear Formula C6H5SCH2C(O)OCH2CH3
Quantity 10 g
Formula Weight 196.27
Percent Purity 97%
Chemical Name or Material Ethyl 2-(phenylthio)acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.