missing translation for 'onlineSavingsMsg'
Learn More

Ethyl (phenylthio)acetate, 97%, Thermo Scientific™

Product Code. 11492948
Change view
Click to view available options
Quantity:
2.5 g
10 g
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11492948 10 g 10g
11482948 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11492948 Supplier Thermo Scientific Alfa Aesar Supplier No. A18664.09
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7605-25-6
Molecular Formula C10H12O2S
Molecular Weight (g/mol) 196.264
MDL Number MFCD00145107
InChI Key SEDRTXNDGKRHBL-UHFFFAOYSA-N
Synonym ethyl phenylthio acetate, ethyl 2-phenylthio acetate, ethyl 2-phenylsulfanyl acetate, acetic acid, phenylthio-, ethyl ester, ethyl 2-phenylthioacetate, phenylthio acetic acid ethyl ester, phenylthio acetic acid, ethyl ester, ethyl phenylsulfanyl acetate, pubchem10853
PubChem CID 97312
IUPAC Name ethyl 2-phenylsulfanylacetate
SMILES CCOC(=O)CSC1=CC=CC=C1

Specifications

Boiling Point 143°C to 145°C (14 mmHg)
Quantity 10 g
Formula Weight 196.27
Percent Purity 97%
Chemical Name or Material Ethyl (phenylthio)acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.