accessibility menu, dialog, popup

UserName

OHFFFF

CAS RN 769-39-1

IUPAC Name: 2,3,5,6-tetrafluorophenol
Synonyms: 2,3,5,6-tetrafluor-phenol
Molecular Weight (g/mol): 166.08
Molecular Formula: C6H2F4O
InChi Key: PBYIIRLNRCVTMQ-UHFFFAOYSA-N
SMILES: OC1=C(F)C(F)=CC(F)=C1F
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)