missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetrafluorophenol, 98%

Product Code. 10782421 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10782421 25 g Glass bottle 25g
10072580 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10782421 Supplier Thermo Scientific Acros Supplier No. 188390250
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 769-39-1 | C6H2F4O | 166.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 769-39-1
Molecular Formula C6H2F4O
Molecular Weight (g/mol) 166.07
MDL Number MFCD00002157
InChI Key PBYIIRLNRCVTMQ-UHFFFAOYSA-N
Synonym phenol, 2,3,5,6-tetrafluoro, unii-tna118i5tg, 2,3,5,6-tetrafluoro phenol, tna118i5tg, 2,3,5,6-tetrafluor-phenol, ccris 5843, zlchem 411, pubchem2135, acmc-1bc5w, 2,3,5,6tetrafluorophenol
PubChem CID 69858
IUPAC Name 2,3,5,6-tetrafluorophenol
SMILES C1=C(C(=C(C(=C1F)F)O)F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 33°C to 38°C
Color White
Boiling Point 140°C
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula HC6F4OH
Quantity 25 g
Formula Weight 166.07
Percent Purity 98%
Physical Form Crystalline Low Melting Solid
Chemical Name or Material 2,3,5,6-Tetrafluorophenol, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2,3,5,6-Tetrafluorophenol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.