missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetrafluorophenol, 97%

Product Code. 11450583
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11450583 25 g 25g
11440583 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11450583 Supplier Thermo Scientific Alfa Aesar Supplier No. A12686.14
 Restricted Item
Estimated Shipment: 23-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 769-39-1 | C6H2F4O | 166.075 g/mol

2,3,5,6-Tetrafluorophenol is used as a cytochrome P450 substrate. 2,3,5,6-Tetrafluorophenol was used in preparation of radioiodinated phenylalanine derivative which is useful in peptide synthesis, technetium-99m labeled antibodies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,5,6-Tetrafluorophenol is used as a cytochrome P450 substrate. 2,3,5,6-Tetrafluorophenol was used in preparation of radioiodinated phenylalanine derivative which is useful in peptide synthesis, technetium-99m labeled antibodies.

Solubility
Partly miscible with water.

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 769-39-1
Molecular Formula C6H2F4O
Molecular Weight (g/mol) 166.075
MDL Number MFCD00002157
InChI Key PBYIIRLNRCVTMQ-UHFFFAOYSA-N
Synonym phenol, 2,3,5,6-tetrafluoro, unii-tna118i5tg, 2,3,5,6-tetrafluoro phenol, tna118i5tg, 2,3,5,6-tetrafluor-phenol, ccris 5843, zlchem 411, pubchem2135, acmc-1bc5w, 2,3,5,6tetrafluorophenol
PubChem CID 69858
IUPAC Name 2,3,5,6-tetrafluorophenol
SMILES C1=C(C(=C(C(=C1F)F)O)F)F

Specifications

Melting Point 28°C to 30°C
Boiling Point 141°C to 143°C
Flash Point 79°C (174°F)
Odor Phenol-like
Quantity 25 g
UN Number UN3261
Beilstein 1911548
Solubility Information Partly miscible with water.
Formula Weight 166.07
Percent Purity 97%
Chemical Name or Material 2,3,5,6-Tetrafluorophenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.