accessibility menu, dialog, popup

UserName

CAS RN 784-04-3

IUPAC Name: 1-(anthracen-9-yl)ethan-1-one
Synonyms: 9-acetylanthracene
Molecular Weight (g/mol): 220.27
Molecular Formula: C16H12O
InChi Key: NXXNVJDXUHMAHU-UHFFFAOYSA-N
SMILES: CC(=O)C1=C2C=CC=CC2=CC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (1)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)