missing translation for 'onlineSavingsMsg'
Learn More

9-Acetylanthracene, 96%

Product Code. 11384524
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11384524 25 g 25g
11374524 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11384524 Supplier Thermo Scientific Alfa Aesar Supplier No. L01278.14
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 784-04-3 | C16H12O | 220.271 g/mol

9-Acetylanthracene is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
9-Acetylanthracene is used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Insoluble in water.

Notes
Store at 15 - 30°C. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 784-04-3
Molecular Formula C16H12O
Molecular Weight (g/mol) 220.271
MDL Number MFCD00001259
InChI Key NXXNVJDXUHMAHU-UHFFFAOYSA-N
Synonym 9-acetylanthracene, 1-anthracen-9-yl ethanone, 9-acetoanthracene, 9-anthryl methyl ketone, ethanone, 1-9-anthracenyl, 1-9-anthryl ethanone, ketone, 9-anthryl methyl, 9-acetyl anthracene, 9-acetylanthrancene, 9-acetylantracene
PubChem CID 69911
IUPAC Name 1-anthracen-9-ylethanone
SMILES CC(=O)C1=C2C=CC=CC2=CC3=CC=CC=C31

Specifications

Melting Point 72°C to 76°C
Quantity 25 g
Beilstein 1370056
Solubility Information Insoluble in water.
Formula Weight 220.27
Percent Purity 96%
Chemical Name or Material 9-Acetylanthracene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.