missing translation for 'onlineSavingsMsg'
Learn More

9-Acetylanthracene, 95%

Product Code. 10606554 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10606554 25 g Glass bottle 25g
10386110 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10606554 Supplier Thermo Scientific Acros Supplier No. 102440250
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 784-04-3 | C16H12O | 220.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 784-04-3
Molecular Formula C16H12O
Molecular Weight (g/mol) 220.27
MDL Number MFCD00001259
InChI Key NXXNVJDXUHMAHU-UHFFFAOYSA-N
Synonym 9-acetylanthracene, 1-anthracen-9-yl ethanone, 9-acetoanthracene, 9-anthryl methyl ketone, ethanone, 1-9-anthracenyl, 1-9-anthryl ethanone, ketone, 9-anthryl methyl, 9-acetyl anthracene, 9-acetylanthrancene, 9-acetylantracene
PubChem CID 69911
IUPAC Name 1-anthracen-9-ylethanone
SMILES CC(=O)C1=C2C=CC=CC2=CC3=CC=CC=C31

Specifikationer

CAS Min % 94.0
CAS Max % 100.0
Melting Point 72°C to 76°C
Color Brown-Yellow to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 25 g
Beilstein 07, II, 450
Formula Weight 220.27
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 9-Acetylanthracene, 95%
Visa mer Visa mindre

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.