accessibility menu, dialog, popup

UserName

FIN

CAS RN 79544-29-9

IUPAC Name: 2-fluoro-6-iodobenzonitrile
Synonyms: 2-fluoro-6-iodobenzonitrile
Molecular Weight (g/mol): 247.01
Molecular Formula: C7H3FIN
InChi Key: FAACTMVXBNSPJA-UHFFFAOYSA-N
SMILES: FC1=CC=CC(I)=C1C#N
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results