missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-iodobenzonitrile, 98%, Thermo Scientific Chemicals

Product Code. 11444643
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11444643 5 g 5g
11434643 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11444643 Supplier Thermo Scientific Alfa Aesar Supplier No. A11576.06
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79544-29-9
Molecular Formula C7H3FIN
Molecular Weight (g/mol) 247.01
MDL Number MFCD00015478
InChI Key FAACTMVXBNSPJA-UHFFFAOYSA-N
Synonym 2-iodo-6-fluorobenzonitrile, 2-cyano-3-fluoro-1-iodobenzene, benzonitrile, 2-fluoro-6-iodo, 6-fluoro-2-iodobenzenecarbonitrile, 2-fluoro-6-iodo-benzonitrile, 6-iodo-2-fluorobenzonitrile, pubchem4780, acmc-209php, ksc494e4h, pharmabridge p-2313
PubChem CID 522722
IUPAC Name 2-fluoro-6-iodobenzonitrile
SMILES FC1=CC=CC(I)=C1C#N

Specifications

Melting Point 48°C to 52°C
Quantity 5 g
UN Number UN3439
Beilstein 4971873
Sensitivity Light sensitive
Formula Weight 247.01
Percent Purity 98%
Chemical Name or Material 2-Fluoro-6-iodobenzonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.