accessibility menu, dialog, popup

UserName

CAS RN 79562-49-5

CH3FNOOF

CAS RN 79562-49-5

IUPAC Name: 1,3-difluoro-2-methyl-4-nitrobenzene
Synonyms: 1,3-difluoro-2-methyl-4-nitrobenzene
Molecular Weight (g/mol): 173.12
Molecular Formula: C7H5F2NO2
InChi Key: AVWNNVJXXMKAPB-UHFFFAOYSA-N
SMILES: CC1=C(F)C=CC(=C1F)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results