CAS RN 82-98-4
CAS RN 82-98-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Piperidolate, MedChemExpress
MedChemExpress Piperidolate is an antimuscarinic, inhibits intestinal cramp induced by acetylcholine (rats and dogs).