accessibility menu, dialog, popup

UserName

CAS RN 83919-23-7

IUPAC Name: (1R,2R,3aS,3bS,9aS,9bR,10S,11aS)-9b-chloro-1-(2-chloroacetyl)-10-hydroxy-2,9a,11a-trimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl furan-2-carboxylate
Synonyms: nasonex mometasone furoate (product) mometasone furoate monohydrate (substance) mometasone preparation nosorex mometasone (substance) danitin rinelon mometasone 17-furoate mometasone furoate monohydrate
Molecular Weight (g/mol): 521.43
Molecular Formula: C27H30Cl2O6
InChi Key: WOFMFGQZHJDGCX-ZULDAHANSA-N
SMILES: C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(Cl)[C@@H](O)C[C@]2(C)[C@@]1(OC(=O)C1=CC=CO1)C(=O)CCl
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (4)
Industry Type 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
  • (1)
  • (4)
  • (4)
  • (1)
  • (1)