accessibility menu, dialog, popup

UserName

H3CCH3OHOH

CAS RN 84-16-2

IUPAC Name: 4-[4-(4-hydroxyphenyl)hexan-3-yl]phenol
Synonyms: hexestrol hexestrol (substance) hexestrol, (r*,r*)-(+-)- hexestrol preparation (product) synoestrolum hexestrol, (r*,s*)- (R-(R*,R*))-hexestrol dihydrodiethylstilbestrol stilbestrol, dihydro- (r*,s*)-hexestrol
Molecular Weight (g/mol): 270.37
Molecular Formula: C18H22O2
InChi Key: PBBGSZCBWVPOOL-UHFFFAOYNA-N
SMILES: CCC(C(CC)C1=CC=C(O)C=C1)C1=CC=C(O)C=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)