missing translation for 'onlineSavingsMsg'
Learn More

7-Chloro-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid (Fluoroquinolonic Acid), Mikromol™

Product Code. 30050651
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050651 25 mg 25mg
17973586 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30050651 Supplier Mikromol Supplier No. MM0018.010025
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

7-Chloro-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid (Fluoroquinolonic Acid)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Fluoroquinolonic acid
CAS 86393-33-1
Analyte or Component Names Fluoroquinolonic acid
Shipping Condition Room Temperature
Molecular Formula C13 H9 Cl F N O3
Certifications/Compliance ISO 17025
Synonym Ciprofloxacin Hydrochloride Imp. A (EP), Ciprofloxacin Imp. A (EP), Enrofloxacin Imp. A (EP), Fluoroquinolonic Acid, 7-Chloro-1-cyclopropyl-6-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid, Ciprofloxacin Hydrochloride Impurity A, Enrofloxacin Impurity A, Ciprofloxacin Impurity A
InChI Formula InChI=1S/C13H9ClFNO3/c14-9-4-11-7(3-10(9)15)12(17)8(13(18)19)5-16(11)6-1-2-6/h3-6H,1-2H2,(H,18,19)
SMILES OC(=O)C1=CN(C2CC2)c3cc(Cl)c(F)cc3C1=O
IUPAC Name 7-chloro-1-cyclopropyl-6-fluoro-4-oxoquinoline-3-carboxylic acid
Molecular Weight (g/mol) 281.67
Quantity 25 mg
Formula Weight 281.0255 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.