accessibility menu, dialog, popup

UserName

CH3NH2Cl

CAS RN 87-60-5

IUPAC Name: 3-chloro-2-methylaniline
Synonyms: 2-methyl-3-chloroaniline
Molecular Weight (g/mol): 141.6
Molecular Formula: C7H8ClN
InChi Key: ZUVPLKVDZNDZCM-UHFFFAOYSA-N
SMILES: CC1=C(N)C=CC=C1Cl
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)