missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-methylaniline, 98+%

Product Code. 11413757
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11413757 250 g 250g
11403757 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11413757 Supplier Thermo Scientific Alfa Aesar Supplier No. A18221.30
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 87-60-5 | C7H8ClN | 141.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-60-5
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.60
MDL Number MFCD00007766
InChI Key ZUVPLKVDZNDZCM-UHFFFAOYSA-N
Synonym 3-chloro-o-toluidine, 2-amino-6-chlorotoluene, benzenamine, 3-chloro-2-methyl, 3-chloro-2-toluidine, scarlet tr base, fast scarlet tr base, 6-chloro-2-aminotoluene, o-toluidine, 3-chloro, 3-chloro-2-methylphenylamine, o-methyl-m-chloroaniline
PubChem CID 6894
IUPAC Name 3-chloro-2-methylaniline
SMILES CC1=C(N)C=CC=C1Cl

Specifications

Melting Point 2°C
Density 1.188
Boiling Point 245°C
Flash Point 113°C (235°F)
Refractive Index 1.589
Quantity 250 g
UN Number UN3429
Beilstein 471697
Formula Weight 141.6
Percent Purity ≥98%
Chemical Name or Material 3-Chloro-2-methylaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.