missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-methylaniline 99%, Thermo Scientific Chemicals

Product Code. 10571174 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10571174 2.5 L Glass bottle 2.5 L
10133290 100 mL Glass bottle 100mL
10543501 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10571174 Supplier Thermo Scientific Acros Supplier No. 146490025
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-60-5
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.60
MDL Number MFCD00007766
InChI Key ZUVPLKVDZNDZCM-UHFFFAOYSA-N
Synonym 3-chloro-o-toluidine, 2-amino-6-chlorotoluene, benzenamine, 3-chloro-2-methyl, 3-chloro-2-toluidine, scarlet tr base, fast scarlet tr base, 6-chloro-2-aminotoluene, o-toluidine, 3-chloro, 3-chloro-2-methylphenylamine, o-methyl-m-chloroaniline
PubChem CID 6894
IUPAC Name 3-chloro-2-methylaniline
SMILES CC1=C(N)C=CC=C1Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Brown-Red or Red to Yellow
Density 1.171g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula ClC6H3(CH3)NH2
Refractive Index 1.5870 to 1.5890 (20°C, 589nm)
Quantity 2.5 L
Beilstein 12, 836
Formula Weight 141.6
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material 3-Chloro-2-methylaniline, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Chloro-2-methylaniline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.