accessibility menu, dialog, popup

UserName

CH3(R)OO(S)CH3(R)CH3CH3(R)CH3O(S)CH3(R)OH(S)CH3CH3OHO(E)(Z)(E)(E)(E)

CAS RN 87081-35-4

IUPAC Name: (2E,5S,6R,7S,9R,10E,12E,15R,16Z,18E)-17-ethyl-6-hydroxy-3,5,7,9,11,15-hexamethyl-19-[(2R,3S)-3-methyl-6-oxo-3,6-dihydro-2H-pyran-2-yl]-8-oxononadeca-2,10,12,16,18-pentaenoic acid
Synonyms: leptomycin
Molecular Weight (g/mol): 540.74
Molecular Formula: C33H48O6
InChi Key: YACHGFWEQXFSBS-NRQIOVFOSA-N
SMILES: CC\C(\C=C\[C@H]1OC(=O)C=C[C@@H]1C)=C\[C@H](C)C\C=C\C(\C)=C\[C@@H](C)C(=O)[C@@H](C)[C@H](O)[C@@H](C)C\C(C)=C\C(O)=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Color 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results