accessibility menu, dialog, popup

UserName

CAS RN 873-76-7

IUPAC Name: (4-chlorophenyl)methanol
Synonyms: P-chlorobenzyl alcohol
Molecular Weight (g/mol): 142.58
Molecular Formula: C7H7ClO
InChi Key: PTHGDVCPCZKZKR-UHFFFAOYSA-N
SMILES: OCC1=CC=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (6)
Percent Purity 
  • (3)
  • (5)
Quantity 
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (6)
  • (3)
  • (5)
  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)