missing translation for 'onlineSavingsMsg'
Learn More

(4-chlorophenyl)methanol, 97%, Thermo Scientific™

Product Code. 10596405
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
Unit Size:
10g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10596405 25 g 25g
10275334 10 g 10g
10062174 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10596405 Supplier Thermo Scientific Maybridge Supplier No. SB01197EB
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 873-76-7
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.582
MDL Number MFCD00004652
InChI Key PTHGDVCPCZKZKR-UHFFFAOYSA-N
Synonym 4-chlorobenzyl alcohol, 4-chlorophenyl methanol, p-chlorobenzyl alcohol, benzenemethanol, 4-chloro, 4-chlorobenzenemethanol, 4-chlorobenzylalcohol, benzyl alcohol, p-chloro, 4-chlorobenzylic alcohol, unii-gf8bs53b6v, ccris 5120
PubChem CID 13397
IUPAC Name (4-chlorophenyl)methanol
SMILES C1=CC(=CC=C1CO)Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Linear Formula ClC6H4CH2OH
Quantity 25 g
Formula Weight 142.58
Percent Purity 97%
Chemical Name or Material (4-Chlorophenyl)methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.