missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzyl alcohol, 99%

Product Code. 11460423
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11460423 100 g 100g
11450423 25 g 25g
11470423 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11460423 Supplier Thermo Scientific Alfa Aesar Supplier No. A11504.22
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 873-76-7 | C7H7ClO | 142.582 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
4-Chlorobenzyl alcohol acts as a reagent for the protection of carboxyl groups as their 4-chlorobenzyl esters, more stable to acid than the corresponding benzyl esters. It is used as solvent in paint stripper and waterborne coatings. It acts as curing agent. It is also used in pharmaceuticals, cosmetics, preservatives, and flavoring & fragrance agents.

Solubility
Soluble in water (2.5 mg/ml at 20°C), and methanol.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 873-76-7
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.582
MDL Number MFCD00004652
InChI Key PTHGDVCPCZKZKR-UHFFFAOYSA-N
Synonym 4-chlorobenzyl alcohol, 4-chlorophenyl methanol, p-chlorobenzyl alcohol, benzenemethanol, 4-chloro, 4-chlorobenzenemethanol, 4-chlorobenzylalcohol, benzyl alcohol, p-chloro, 4-chlorobenzylic alcohol, unii-gf8bs53b6v, ccris 5120
PubChem CID 13397
IUPAC Name (4-chlorophenyl)methanol
SMILES C1=CC(=CC=C1CO)Cl

Specifications

Melting Point 68°C to 72°C
Boiling Point 234°C
Quantity 100 g
Beilstein 636502
Solubility Information Soluble in water (2.5mg/ml at 20°C),and methanol.
Formula Weight 142.59
Percent Purity 99%
Chemical Name or Material 4-Chlorobenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.