accessibility menu, dialog, popup

UserName

CAS RN 90-30-2

IUPAC Name: N-phenylnaphthalen-1-amine
Synonyms: PANA fenyl-α-naftylamin antigene pan antioxidant pan neozone A amoco 32 1-N-phenylnaphthylamine nonox A aceto pan N-fenyl-1-aminonaftalen
Molecular Weight (g/mol): 219.29
Molecular Formula: C16H13N
InChi Key: XQVWYOYUZDUNRW-UHFFFAOYSA-N
SMILES: N(C1=CC=CC=C1)C1=C2C=CC=CC2=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)