accessibility menu, dialog, popup

UserName

HOOHN

CAS RN 91-40-7

IUPAC Name: 2-(phenylamino)benzoic acid
Synonyms: phenylanthranilic acid 2-carboxydiphenylamine fenamic acid dpc cpd fenamic acid, ca salt (2:1) monosodium salt fenamic acid N-phenyl-o-aminobenzoic acid ca salt (2:1) fenamic acid diphenylamine-2-carboxylate fenamic acid, monosodium salt
Molecular Weight (g/mol): 213.24
Molecular Formula: C13H11NO2
InChi Key: ZWJINEZUASEZBH-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC=C1NC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
  • (3)
Quantity 
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (5)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)