accessibility menu, dialog, popup

UserName

CAS RN 98-91-9

IUPAC Name: benzenecarbothioic S-acid
Synonyms: benzenecarbothioic acid
Molecular Weight (g/mol): 138.18
Molecular Formula: C7H6OS
InChi Key: UIJGNTRUPZPVNG-UHFFFAOYSA-N
SMILES: SC(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results