missing translation for 'onlineSavingsMsg'
Learn More

Thiobenzoic acid, 90%

Product Code. 10616152 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity Packaging Conditionnement
10616152 100 g Glass bottle 100g
10317120 25 g Glass bottle 25g
10381334 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 10616152 Fournisseur Thermo Scientific Acros Code fournisseur 138681000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Voir les produits de remplacement

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 98-91-9 | C7H6OS | 138.184 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 98-91-9
MDL Number MFCD00004852
InChI Key UIJGNTRUPZPVNG-UHFFFAOYSA-N
Synonym thiobenzoic acid, benzenecarbothioic acid, benzoyl thiol, monothiobenzoic acid, benzoic acid, thio, acido mercaptobenzoico, unii-gbg5rlo56n, ccris 8913, acido mercaptobenzoico italian, gbg5rlo56n
PubChem CID 7414
IUPAC Name benzenecarbothioic S-acid
SMILES C1=CC=C(C=C1)C(=O)S

Spécification

Melting Point 15.0°C to 18.0°C
Color Yellow to Brown
Density 1.1700g/mL
Boiling Point 122.0°C (30.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Molecular Formula C7H6OS
Linear Formula C6H5COSH
Refractive Index 1.6020 to 1.6080
Quantity 100 g
Beilstein 09, 419
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with organic solvents
Molecular Weight (g/mol) 138.184
Formula Weight 138.18
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material Thiobenzoic acid
Afficher plus Afficher moins
compliance-icons
Identifiant produit
  • Thiobenzoic acid
Mention d'avertissement
  • Warning
Catégorie de danger
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Conseils de prudence
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.