accessibility menu, dialog, popup

UserName

CH3H2NNOO

CAS RN 99-55-8

IUPAC Name: 2-methyl-5-nitroaniline
Synonyms: 2-amino-4-nitrotoluene
Molecular Weight (g/mol): 152.15
Molecular Formula: C7H8N2O2
InChi Key: DSBIJCMXAIKKKI-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1N)[N+]([O-])=O
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results