missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-nitroaniline, 96%, Thermo Scientific™

Product Code. 10724943 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10724943 500 g 500g
10653763 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10724943 Supplier Thermo Scientific Acros Supplier No. 364915000
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-55-8
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.15
MDL Number MFCD00007741
InChI Key DSBIJCMXAIKKKI-UHFFFAOYSA-N
Synonym 2-amino-4-nitrotoluene, 5-nitro-o-toluidine, fast scarlet g, pnot, scarlet g base, 2-methyl-5-nitrobenzenamine, 4-nitro-2-aminotoluene, devol scarlet b, diabase scarlet g, scarlet base nsp
PubChem CID 7444
ChEBI CHEBI:66891
IUPAC Name 2-methyl-5-nitroaniline
SMILES CC1=CC=C(C=C1N)[N+]([O-])=O

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 104.0°C to 107.0°C
Color Ochre to Orange-Yellow
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Linear Formula CH3C6H3(NO2)NH2
Quantity 500 g
Beilstein 12, 844
Solubility Information Solubility in water: insoluble.
Formula Weight 152.15
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-5-nitroaniline, 96%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Methyl-5-nitroaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 2
Hazard Statement
  • H351-Suspected of causing cancer.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.