missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
HIST1H2AB Polyclonal antibody specifically detects HIST1H2AB in Human samples. It is validated for Western Blot
Specifications
Specifications
| Applications | Western Blot |
| Conjugate | PE |
| Gene Alias | histone H2A type 1-B/E, H2A/m, H2AFM, histone cluster 1, H2ab |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human HIST1H2AB. |
| Purification Method | Antigen and protein A Affinity-purified |
| Quantity | 0.1 mL |
| Primary or Secondary | Primary |
| Gene ID (Entrez) | 8335 |
| Content And Storage | Store at 4C in the dark. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?