missing translation for 'onlineSavingsMsg'
Learn More

1,1,1,3,3,3-Hexafluoropropan-2-ol, Mikromol™

Product Code. 17933836
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17933836 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17933836 Supplier Mikromol Supplier No. MM3315.010100
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1,1,1,3,3,3-Hexafluoropropan-2-ol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,1,1,3,3,3-hexafluoro-2-propanol
CAS 920-66-1
Analyte or Component Names 1,1,1,3,3,3-hexafluoro-2-propanol
Shipping Condition Room Temperature
Molecular Formula C3 H2 F6 O
Certifications/Compliance ISO 9001
Synonym 2-Propanol, 1,1,1,3,3,3-hexafluoro-, 1,1,1,3,3,3-Hexafluoro-2-hydroxypropane, 1,1,1,3,3,3-Hexafluoroisopropanol, 1,1,1,3,3,3-Hexafluoroisopropyl alcohol, 1,1,1,3,3,3-Hexafluoropropane-2-ol, 1,1,1,3,3,3-Hexafluoropropanol, 2,2,2-Trifluoro-1-(trifluoromethyl)ethanol, Bis(trifluoromethyl)methanol, HFIP, Hexafluoroisopropanol, Hexafluoroisopropyl alcohol, NSC 96336, Sevoflurane Related Compound, Sevoflurane Impurity C
InChI Formula InChI=1S/C3H2F6O/c4-2(5,6)1(10)3(7,8)9/h1,10H
SMILES OC(C(F)(F)F)C(F)(F)F
IUPAC Name 1,1,1,3,3,3-hexafluoropropan-2-ol
Molecular Weight (g/mol) 168.04
Quantity 100 mg
Formula Weight 168.001 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.