missing translation for 'onlineSavingsMsg'
Learn More

1,1′,1′′-(Methoxymethanetriyl)tribenzene (Methyl Trityl Ether), Mikromol™

Product Code. 17940626
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17940626 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17940626 Supplier Mikromol Supplier No. MM0173.100025
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1,1′,1′′-(Methoxymethanetriyl)tribenzene (Methyl Trityl Ether)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Methyl Triphenylmethyl Ether
CAS 596-31-6
Analyte or Component Names Methyl Triphenylmethyl Ether
Shipping Condition Ice pack (-20°C)
Molecular Formula C20 H18 O
Certifications/Compliance ISO 9001
Synonym 1,1',1'-(Methoxymethylidyne)tris-benzene, Ether, methyl trityl (6CI,7CI,8CI), 1,1',1'-(Methoxymethylidyne)tris[benzene], Methoxytriphenylmethane, Methyl triphenylmethyl ether, Methyl trityl ether, NSC 64, Triphenylmethoxymethane, Triphenylmethyl methyl ether, Trityl methyl ether, Zidovudine Impurity K
InChI Formula InChI=1S/C20H18O/c1-21-20(17-11-5-2-6-12-17,18-13-7-3-8-14-18)19-15-9-4-10-16-19/h2-16H,1H3
SMILES COC(c1ccccc1)(c2ccccc2)c3ccccc3
IUPAC Name [methoxy(diphenyl)methyl]benzene
Molecular Weight (g/mol) 274.36
Quantity 25 mg
Formula Weight 274.1358 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.