missing translation for 'onlineSavingsMsg'
Learn More

1,2,3,4-Tetrahydro-4,8-dihydroxy-2-methyl-isoquinoline Hydrochloride, Mikromol™

Product Code. 14871351
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
14871351 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 14871351 Supplier Mikromol Supplier No. MM0081.06
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1,2,3,4-Tetrahydro-4,8-dihydroxy-2-methyl-isoquinoline Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2,3,4-Tetrahydro-4,8-dihydroxy-2-methyl-isoquinoline Hydrochloride
CAS 2514676-44-7
Analyte or Component Names 1,2,3,4-Tetrahydro-4,8-dihydroxy-2-methyl-isoquinoline Hydrochloride
Shipping Condition Ice pack (-20°C)
Molecular Formula C10 H13 N O2 . Cl H
Certifications/Compliance ISO 17025
Synonym 1,2,3,4-Tetrahydro-4,8-dihydroxy-2-methyl-isoquinoline Hydrochloride
InChI Formula InChI=1S/C10H13NO2.ClH/c1-11-5-8-7(10(13)6-11)3-2-4-9(8)12;/h2-4,10,12-13H,5-6H2,1H3;1H
SMILES Cl.CN1CC(O)c2cccc(O)c2C1
IUPAC Name 2-methyl-3,4-dihydro-1H-isoquinoline-4,8-diol;hydrochloride
Molecular Weight (g/mol) 215.68
Quantity 50 mg
Formula Weight 215.0713 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.