missing translation for 'onlineSavingsMsg'
Learn More

1-[2-Amino-1-(4-methoxyphenyl)ethyl]cyclohexanol Hydrochloride, TRC

Product Code. 30134600
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134600 10 g 10g
30135992 25 g 25g
17962485 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134600 Supplier Toronto Research Chemicals Supplier No. TRCA61196810G
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 130198-05-9
Molecular Formula C15 H23 N O2 . Cl H
Molecular Weight (g/mol) 285.81
Synonym 1-[(1RS)-2-Amino-1-(4-methoxyphenyl)ethyl]cyclohexanol Hydrochloride, Venlafaxine Hydrochloride Imp. C (EP) as Hydrochloride, 1-[2-Amino-1-(4-methoxyphenyl)ethyl]-cyclohexanol Hydrochloride, Cyclohexanol, 1-[2-amino-1-(4-methoxyphenyl)ethyl]-, hydrochloride (9CI), Cyclohexanol, 1-[2-amino-1-(4-methoxyphenyl)ethyl]-, hydrochloride, (±)-, 1-[2-Amino-1-(4-methoxyphenyl)ethyl]cyclohexano Hydrochloride l, 1-[2-Amino-1-(p-methoxyphenyl)ethyl]cyclohexanol Hydrochloride, N,N-Didesmethylvenlafaxine Hydrochloride
IUPAC Name 1-[2-amino-1-(4-methoxyphenyl)ethyl]cyclohexan-1-ol;hydrochloride
SMILES Cl.COc1ccc(cc1)C(CN)C2(O)CCCCC2

Specifications

Analyte or Component Names 1-[(1RS)-2-Amino-1-(4-methoxyphenyl)ethyl]cyclohexanol Hydrochloride
Shipping Condition Room Temperature
InChI Formula InChI=1S/C15H23NO2.ClH/c1-18-13-7-5-12(6-8-13)14(11-16)15(17)9-3-2-4-10-15;/h5-8,14,17H,2-4,9-11,16H2,1H3;1H
Solution Type Neat
Industry Type Pharmaceutical
Quantity 10 g
Formula Weight 285.1496
Chemical Name or Material 1-[(1RS)-2-Amino-1-(4-methoxyphenyl)ethyl]cyclohexanol Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.