missing translation for 'onlineSavingsMsg'
Learn More

1,2-Benzisothiazol-3(2H)-one, Mikromol™

Product Code. 30049933
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30049933 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30049933 Supplier Mikromol Supplier No. MM1273.110025
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1,2-Benzisothiazol-3(2H)-one
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2-Benzisothiazol-3(2H)-one
CAS 2634-33-5
Analyte or Component Names 1,2-Benzisothiazol-3(2H)-one
Shipping Condition Room Temperature
Molecular Formula C7 H5 N O S
Certifications/Compliance ISO 9001
Synonym 1,2-Benzisothiazol-3(2H)-one, 1,2-Benzisothiazolin-3-one (6CI,7CI,8CI), 1,2-Benzisothiazol-3-one, 1,2-Benzisothiazolone, 1,2-Benzoisothiazol-3-one, 2,3-Dihydrobenzisothiazol-3-one, 3-Hydroxy-1,2-benzisothiazole, AQ, AQ (antibacterial), Acticide B 20N, Acticide BIT, Acticide BW 20, Apizas AP-DS, BIT, BIT 10W, BIT 20, Benzisothiazolin-3-one, Benzisothiazolinone, Benzisothiazolone, Benzo[d]isothiazol-3(2H)-one, Benzocil, Benzoisothiazol-3-one, Bestcide 200K, Bioban BIT 20DPG, Bioban Ultra Bit, Canguard BIT, Canguard BIT 20AS-E, Canguard BIT 20DPG, Canguard Ultra BIT 20LE, Denicide BIT, Denicide BIT 20N, GXL, Koralone B 119, Mergal 753, Nipacide BIT, Nipacide BIT 10W, Nipacide BIT 20, Nuosept 485, Nuosept 491, Nuosept 495, Parmetol B 70, Parmetol D 11, Preventol BIT 20D, Proxel AB, Proxel AQ, Proxel BD, Proxel BD 20, Proxel BDN, Proxel CF, Proxel GXL, Proxel GXL(S), Proxel HL 2, Proxel LV, Proxel LV-S, Proxel PL, Proxel Press Paste, Proxel Press Paste D, Proxel TN, Proxel Ultra 5, Proxel XL, Proxel XL 2, Rocima 640, SD 202, SD 202 (bactericide), San-aibac AP, Topcide 600, Troysan 1050, XBINX
InChI Formula InChI=1S/C7H5NOS/c9-7-5-3-1-2-4-6(5)10-8-7/h1-4H,(H,8,9)
SMILES O=C1NSc2ccccc12
IUPAC Name 1,2-benzothiazol-3-one
Molecular Weight (g/mol) 151.19
Quantity 25 mg
Formula Weight 151.0092 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.