missing translation for 'onlineSavingsMsg'
Learn More

1,2-Benzisoxazol-3-ylacetic Acid, Mikromol™

Product Code. 17952416
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17952416 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17952416 Supplier Mikromol Supplier No. MM1379.040025
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1,2-Benzisoxazol-3-ylacetic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,2-Benzisoxazol-3-ylacetic Acid
CAS 4865-84-3
Analyte or Component Names 1,2-Benzisoxazol-3-ylacetic Acid
Shipping Condition Room Temperature
Molecular Formula C9 H7 N O3
Certifications/Compliance ISO 17025
Synonym 1,2-Benzisoxazole-3-acetic acid, (1,2-Benzoxazol-3-yl)acetic acid, (Benzo[d]isoxazol-3-yl)acetic acid, 1,2-Benzisoxazol-3-ylacetic acid, 2-(Benzisoxazol-3-yl)acetic acid, 2-(Benzo[d]isoxazol-3-yl)acetic acid, Benzisoxazol-3-ylacetic acid, NSC 179803
InChI Formula InChI=1S/C9H7NO3/c11-9(12)5-7-6-3-1-2-4-8(6)13-10-7/h1-4H,5H2,(H,11,12)
SMILES OC(=O)Cc1noc2ccccc12
IUPAC Name 2-(1,2-benzoxazol-3-yl)acetic acid
Molecular Weight (g/mol) 177.16
Quantity 25 mg
Formula Weight 177.0426 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.