missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Fluorophenyl)propanol, 95%, Thermo Scientific™

Product Code. 15404587
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15404587 10 g 10g
15414587 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15404587 Supplier Thermo Scientific Alfa Aesar Supplier No. H31929.09
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as an intermediate in organic synthesis and is a pharmaceutical intermediate.

Notes
Store in cool dry place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156022-15-0
Molecular Formula C9H11FO
Molecular Weight (g/mol) 154.184
MDL Number MFCD08062368
InChI Key MCENMZBHIHBHPB-UHFFFAOYSA-N
Synonym 1-2-fluorophenyl propan-1-ol, 1-2-fluorophenyl propanol, 2-fluorophenylpropanol, pubchem19915, 1-2-fluorophenyl-1-propanol, benzenemethanol, alpha-ethyl-2-fluoro-9ci
PubChem CID 21994116
IUPAC Name 1-(2-fluorophenyl)propan-1-ol
SMILES CCC(C1=CC=CC=C1F)O

Specifications

Quantity 10 g
Formula Weight 154.18
Percent Purity 95%
Chemical Name or Material 1-(2-Fluorophenyl)propanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.