missing translation for 'onlineSavingsMsg'
Learn More

1-(2-Methoxyphenyl)tricyclo[3.3.1.13,7]decane, Mikromol™

Product Code. 30050779
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050779 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30050779 Supplier Mikromol Supplier No. MM0681.01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1-(2-Methoxyphenyl)tricyclo[3.3.1.13,7]decane
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-(2-Methoxyphenyl)tricyclo[3.3.1.13,7]decane
CAS 43109-77-9
Analyte or Component Names 1-(2-Methoxyphenyl)tricyclo[3.3.1.13,7]decane
Shipping Condition Room Temperature
Molecular Formula C17 H22 O
Certifications/Compliance ISO 17025
Synonym Adapalene Imp. C (EP), Adapalene USP Related Compound C, Adapalene USP RC C, 1-(2-Methoxyphenyl)tricyclo[3.3.1.13,7]decane, Adapalene Related Compound C, Adapalene Impurity C
InChI Formula InChI=1S/C17H22O/c1-18-16-5-3-2-4-15(16)17-9-12-6-13(10-17)8-14(7-12)11-17/h2-5,12-14H,6-11H2,1H3
SMILES COc1ccccc1C23CC4CC(CC(C4)C2)C3
IUPAC Name 1-(2-methoxyphenyl)adamantane
Molecular Weight (g/mol) 242.36
Quantity 50 mg
Formula Weight 242.1671 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.