missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-[(2RS)-2-(2,4-Dichlorophenyl)-2-[(thiophen-3-yl)methoxy]ethyl]-1H-imidazole Hydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
1-[(2RS)-2-(2,4-Dichlorophenyl)-2-[(thiophen-3-yl)methoxy]ethyl]-1H-imidazole Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Tioconazole Related Compound A |
| CAS | 61709-33-9 |
| Analyte or Component Names | Tioconazole Related Compound A |
| Shipping Condition | Room Temperature |
| Molecular Formula | C16 H14 Cl2 N2 O S . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1H-Imidazole, 1-[2-(2,4-dichlorophenyl)-2-(3-thienylmethoxy)ethyl]-, hydrochloride (1:1), 1H-Imidazole, 1-[2-(2,4-dichlorophenyl)-2-(3-thienylmethoxy)ethyl]-, monohydrochloride (9CI), Tioconazole Imp. A (EP), Tioconazole USP Related Compound A, Tioconazole USP RC A, Tioconazole Related Compound A as Hydrochloride, Tioconazole Impurity A as Hydrochloride |
| InChI Formula | InChI=1S/C16H14Cl2N2OS.ClH/c17-13-1-2-14(15(18)7-13)16(8-20-5-4-19-11-20)21-9-12-3-6-22-10-12;/h1-7,10-11,16H,8-9H2;1H |
| SMILES | Cl.Clc1ccc(C(Cn2ccnc2)OCc3ccsc3)c(Cl)c1 |
| IUPAC Name | 1-[2-(2,4-dichlorophenyl)-2-(thiophen-3-ylmethoxy)ethyl]imidazole;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction