missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-trichloro-2-iodobenzene, Thermo Scientific™

Product Code. 10174194
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10174194 1 g 1g
10682505 5 g 5g
10555645 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10174194 Supplier Thermo Scientific Maybridge Supplier No. TL00270DA
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 6324-50-1
Molecular Formula C6H2Cl3I
Molecular Weight (g/mol) 307.336
InChI Key XXFFSBRDPQFIPO-UHFFFAOYSA-N
PubChem CID 232054
IUPAC Name 1,3,5-trichloro-2-iodobenzene
SMILES C1=C(C=C(C(=C1Cl)I)Cl)Cl

Specifications

Assay Percent Range 97%
Quantity 1 g
Synonym 2,4,6-trichloroiodobenzene, 1-iodo-2,4,6-trichlorobenzene, benzene,1,3,5-trichloro-2-iodo, benzene, 1,3,5-trichloro-2-iodo, acmc-20aof6, 2,4,6-trichloro-1-iodobenzene, 2-iodo-1,3,5-trichlorobenzene, 1,3,5-trichloro-2-iodo-benzene, 1,3,5-tris chloranyl-2-iodanyl-benzene
Formula Weight 307.35
Percent Purity 97%
Chemical Name or Material 1, 3, 5-Trichloro-2-iodobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.