missing translation for 'onlineSavingsMsg'
Learn More

1,3,5-Trimethoxybenzene, Mikromol™

Product Code. 30458585
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30458585 4 x 25 mg -
30051577 100 mg 100mg
30051933 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30458585 Supplier Mikromol Supplier No. MM0066.0500254AMP
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1,3,5-Trimethoxybenzene, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,3,5-Trimethoxybenzene
CAS 621-23-8
Analyte or Component Names 1,3,5-Trimethoxybenzene
Shipping Condition Room Temperature
Molecular Formula C9 H12 O3
Certifications/Compliance ISO 17025
Synonym 1,3,5-Trimethoxybenzene
InChI Formula InChI=1S/C9H12O3/c1-10-7-4-8(11-2)6-9(5-7)12-3/h4-6H,1-3H3
Solution Type Neat
SMILES COc1cc(OC)cc(OC)c1
IUPAC Name 1,3,5-trimethoxybenzene
Molecular Weight (g/mol) 168.19
Quantity 4 x 25 mg
Formula Weight 168.0786 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.