missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(3-Chlorophenyl)piperazine Hydrochloride, Mikromol
Description
1-(3-Chlorophenyl)piperazine Hydrochloride, Mikromol
Specifications
Specifications
| Chemical Name or Material | 1-(3-Chlorophenyl) piperazine (hydrochloride) |
| CAS | 13078-15-4 |
| Analyte or Component Names | 1-(3-Chlorophenyl) piperazine (hydrochloride) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H13 Cl N2 . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | 1-(3-Chlorophenyl)piperazine Monohydrochloride, 1-(m-Chlorophenyl)piperazine Monohydrochloride, 1-(3-Chlorophenyl)piperazine Hydrochloride, 1-(m-Chlorophenyl)piperazine hydrochloride, N-(3-Chlorophenyl)piperazine hydrochloride, m-CPP hydrochloride |
| InChI Formula | InChI=1S/C10H13ClN2.ClH/c11-9-2-1-3-10(8-9)13-6-4-12-5-7-13;/h1-3,8,12H,4-7H2;1H |
| Solution Type | Neat |
| SMILES | Cl.Clc1cccc(c1)N2CCNCC2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?