missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dichloro-5-(chloromethyl)benzene, 97%, Thermo Scientific™

Product Code. p-4510515
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10501133 1 g Amber Glass Bottle 1g
10672463 10 g Amber Glass Bottle 10g
10357684 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10501133 Supplier Thermo Scientific Maybridge Supplier No. SPB00444DA
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 3290-06-0
Molecular Formula C7H5Cl3
Molecular Weight (g/mol) 195.467
InChI Key ZFLRKAMKGYNFPH-UHFFFAOYSA-N
Synonym 3,5-dichlorobenzyl chloride, 1,3-dichloro-5-chloromethyl benzene, benzene, 1,3-dichloro-5-chloromethyl, 3,5-dichlorobenzylchloride, unii-c076i19eql, 5-chloromethyl-1,3-dichlorobenzene, pubchem16695, alpha,3,5-trichlorotoluene, 3,4-dichloro benzyl chloride, ksc495g8l
PubChem CID 137880
IUPAC Name 1,3-dichloro-5-(chloromethyl)benzene
SMILES C1=C(C=C(C=C1Cl)Cl)CCl

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula Cl2C6H3CH2Cl
Quantity 1 g
Formula Weight 195.48
Percent Purity 97%
Chemical Name or Material 1, 3-Dichloro-5-(chloromethyl)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.