missing translation for 'onlineSavingsMsg'
Learn More

1,3-Dihydroxybenzene (Resorcinol), Mikromol™

Product Code. 18013960
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18013960 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18013960 Supplier Mikromol Supplier No. MM0294.010250
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1,3-Dihydroxybenzene (Resorcinol)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Resorcinol (1,3-Benzenediol)
CAS 108-46-3
Analyte or Component Names Resorcinol (1,3-Benzenediol)
Shipping Condition Room Temperature
Molecular Formula C6 H6 O2
Certifications/Compliance ISO 17034
Synonym Fluorescein Sodium Imp. A (EP), Fluorescein Imp. A (EP), Hexylresorcinol Imp. B (EP), Hymecromone Imp. A (EP), Resorcinol, 1,3-Dihydroxybenzene, 1,3-Benzenediol, Resorcinol (8CI), 1,3-Phenylenediol, 3-Hydroxyphenol, C.I. 76505, C.I. Developer 4, C.I. Oxidation Base 31, Developer O, Developer R, Developer RS, Durafur Developer G, Fouramine RS, Fourrine 79, Fourrine EW, NSC 1571, Nako TGG, Pelagol Grey RS, Pelagol RS, RS 11H, RS 11L, Redimix 401RAP60, Reso, Resorcin, Resorcinol 80, Rezorsine, Rodol RS, m-Benzenediol, m-Dihydroxybenzene, m-Hydroquinone, m-Hydroxyphenol, m-Phenylenediol, m-Resorcinol, Fluorescein Sodium Impurity A, Hexylresorcinol Impurity B, Hymecromone Impurity A, Fluorescein Impurity A
InChI Formula InChI=1S/C6H6O2/c7-5-2-1-3-6(8)4-5/h1-4,7-8H
SMILES Oc1cccc(O)c1
IUPAC Name benzene-1,3-diol
Molecular Weight (g/mol) 110.11
Quantity 250 mg
Formula Weight 110.0368 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.