missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Fluorobenzyl)piperazine, 97%

Product Code. 15431508
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15431508 1 g 1g
15441508 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15431508 Supplier Thermo Scientific Alfa Aesar Supplier No. H55295.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 55513-19-4 | C11H15FN2 | 194.253 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55513-19-4
Molecular Formula C11H15FN2
Molecular Weight (g/mol) 194.253
MDL Number MFCD02242870
InChI Key ITHBJSRWFNLKIH-UHFFFAOYSA-N
Synonym 1-3-fluorobenzyl piperazine, 1-3-fluorophenyl methyl piperazine, 1-3-fluoro-benzyl-piperazine, 3-fluorophenyl methyl piperazine, 1-3-fluorobenz-yl piperazine, piperazine, 1-3-fluorophenyl methyl, 1-3-fluorophenyl methyl piperazine trifluoroacetate
PubChem CID 903813
IUPAC Name 1-[(3-fluorophenyl)methyl]piperazine
SMILES C1CN(CCN1)CC2=CC(=CC=C2)F

Specifications

Color Colorless to Orange
Density 1.106 g/mL
Boiling Point 281°C
Flash Point >110°C (230°F)
Refractive Index 1.529
Quantity 1 g
Formula Weight 194.25
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-(3-Fluorobenzyl)piperazine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.