missing translation for 'onlineSavingsMsg'
Learn More

1,4-Bis[(4-chlorophenyl)phenylmethyl]piperazine, Mikromol™

Product Code. 17933646
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17933646 25 mg 25mg
17904266 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17933646 Supplier Mikromol Supplier No. MM0380.040025
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

1,4-Bis[(4-chlorophenyl)phenylmethyl]piperazine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,4-Bis[(4-chlorophenyl)phenylmethyl]piperazine
CAS 346451-15-8
Analyte or Component Names 1,4-Bis[(4-chlorophenyl)phenylmethyl]piperazine
Shipping Condition Room Temperature
Molecular Formula C30 H28 Cl2 N2
Certifications/Compliance ISO 17025
Synonym Cetirizine Dihydrochloride Imp. D (EP), Cetirizine Imp. D (EP), Meclozine Hydrochloride Imp. F (EP), Meclozine Imp. F (EP), 1,4-Bis[(4-chlorophenyl)phenylmethyl]piperazine, 1,4-Bis(4-chlorobenzhydryl)piperazine, Buclizine Hydrochloride Imp. A (BP), Cetirizine Dihydrochloride Impurity D, Meclozine Dihydrochloride Impurity F, Cetirizine Impurity D, Meclozine Impurity F
InChI Formula InChI=1S/C30H28Cl2N2/c31-27-15-11-25(12-16-27)29(23-7-3-1-4-8-23)33-19-21-34(22-20-33)30(24-9-5-2-6-10-24)26-13-17-28(32)18-14-26/h1-18,29-30H,19-22H2
SMILES Clc1ccc(cc1)C(N2CCN(CC2)C(c3ccccc3)c4ccc(Cl)cc4)c5ccccc5
IUPAC Name 1,4-bis[(4-chlorophenyl)-phenylmethyl]piperazine
Molecular Weight (g/mol) 487.46
Quantity 25 mg
Formula Weight 486.163 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.