missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,8-Dihydroxyanthracene-9,10-dione (Danthron), Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
1,8-Dihydroxyanthracene-9,10-dione (Danthron)
Specifications
Specifications
| Chemical Name or Material | Dantron |
| CAS | 117-10-2 |
| Analyte or Component Names | Dantron |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H8 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1,8-Dihydroxyanthracene-9,10-dione, Dithranol Imp. B (EP), Danthron, 1,8-Dihydroxy-9,10-anthraquinone, 1,8-Dihydroxyanthraquinone, 1,8-Dioxyanthraquinone, Altan, Antrapurol, Chrysazin, Danthrone, Dantron, Diaquone, Dionone, Dorbane, Istin, Istizin, Laxanorm, Laxanthreen, Laxipur, Laxipurin, Modane, NSC 38626, NSC 646568, NSC 7210, Zwitsalax, Dithranol Impurity B |
| InChI Formula | InChI=1S/C14H8O4/c15-9-5-1-3-7-11(9)14(18)12-8(13(7)17)4-2-6-10(12)16/h1-6,15-16H |
| SMILES | Oc1cccc2C(=O)c3cccc(O)c3C(=O)c12 |
| IUPAC Name | 1,8-dihydroxyanthracene-9,10-dione |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction