missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-3,5-difluorobenzene, 98%

Product Code. 11427966
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
250 g
Unit Size:
100g
10g
250g
25g
5g
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11427966 5 g 5g
11404200 10 g 10g
11437966 25 g 25g
11447966 100 g 100g
11414200 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11427966 Supplier Thermo Scientific Alfa Aesar Supplier No. A14053.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 461-96-1 | C6H3BrF2 | 192.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 461-96-1
Molecular Formula C6H3BrF2
Molecular Weight (g/mol) 192.99
MDL Number MFCD00010305
InChI Key JHLKSIOJYMGSMB-UHFFFAOYSA-N
Synonym 3,5-difluorobromobenzene, 1-bromo-3,5-difluoro-benzene, 1-bromo-3,5-difluoro benzene, benzene, 1-bromo-3,5-difluoro, 3,5-difluoro-bromobenzene, pubchem1042, acmc-1ae0k, 3,5-difluoro bromobenzene, bromo-3,5-difluorobenzene, 3,5-difluorophenyl bromide
PubChem CID 136313
IUPAC Name 1-bromo-3,5-difluorobenzene
SMILES FC1=CC(Br)=CC(F)=C1

Specifications

Melting Point -27°C
Density 1.676
Boiling Point 140°C to 141°C
Flash Point 44°C (111°F)
Refractive Index 1.499
Quantity 5 g
UN Number UN1993
Beilstein 1931520
Formula Weight 192.99
Percent Purity 98%
Chemical Name or Material 1-Bromo-3,5-difluorobenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.