missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-n-heptylbenzene, 97%, Thermo Scientific Chemicals

Product Code. 11464923
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11464923

Brand: Thermo Scientific Alfa Aesar A12235.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76287-49-5
Molecular Formula C13H19Br
Molecular Weight (g/mol) 255.199
MDL Number MFCD00061115
InChI Key YHKMTIJHJWYYAG-UHFFFAOYSA-N
Synonym 1-bromo-4-n-heptylbenzene, 1-4-bromophenyl heptane, 4-heptylbromobenzene, benzene, 1-bromo-4-heptyl, 4-bromo-1-heptylbenzene, 1-bromo-4-heptyl-benzene, acmc-209p2u, 1-bromanyl-4-heptyl-benzene, ksc912e9b
PubChem CID 2775100
IUPAC Name 1-bromo-4-heptylbenzene
SMILES CCCCCCCC1=CC=C(C=C1)Br

Specifications

Quantity 25 g
Formula Weight 255.2
Percent Purity 97%
Chemical Name or Material 1-Bromo-4-n-heptylbenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.