missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-(trifluoromethylthio)benzene, 97%

Product Code. 11459896
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
11459896 1 g 1g
11469896 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 11459896

Brand: Thermo Scientific Alfa Aesar B20874.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 333-47-1 | C7H4BrF3S | 257.068 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 333-47-1
Molecular Formula C7H4BrF3S
Molecular Weight (g/mol) 257.068
MDL Number MFCD00040840
InChI Key CQQBVMXLTBXNLB-UHFFFAOYSA-N
Synonym 4-trifluoromethylthio bromobenzene, 1-bromo-4-trifluoromethyl sulfanyl benzene, 4-bromophenyl trifluoromethyl sulfide, 4-bromophenyl trifluoromethyl sulphide, 1-bromo-4-trifluoromethylthio benzene, 1-bromo-4-trifluoromethylthiobenzene, 4-bromophenyl trifluoromethyl sulfane, 1-bromo-4-trifluoromethyl thio benzene, 4-bromo-1-trifluoromethylthio benzene, 1-bromo-4-trifluoromethylsulfanyl benzene
PubChem CID 736413
IUPAC Name 1-bromo-4-(trifluoromethylsulfanyl)benzene
SMILES C1=CC(=CC=C1SC(F)(F)F)Br

Specifications

Density 1.71
Boiling Point 199°C to 200°C
Flash Point 83°C (181°F)
Refractive Index 1.517
Quantity 1 g
Beilstein 2361926
Formula Weight 257.07
Percent Purity 97%
Chemical Name or Material 1-Bromo-4-(trifluoromethylthio)benzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.